missing translation for 'onlineSavingsMsg'
Learn More

3-Butoxyphenol 96.0+%, TCI America™

Catalog No. B07015G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B07015G 5 g
B070125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B07015G Supplier TCI America Supplier No. B07015G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 18979-72-1
Molecular Formula C10H14O2
Molecular Weight (g/mol) 166.22
MDL Number MFCD00058976
InChI Key VGIJZDWQVCXVNL-UHFFFAOYSA-N
Synonym Resorcinol Monobutyl Ether
PubChem CID 87876
IUPAC Name 3-butoxyphenol
SMILES CCCCOC1=CC=CC(=C1)O

Specifications

Color Yellow
Boiling Point 269°C
Quantity 5 g
Formula Weight 166.22
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Butoxyphenol