accessibility menu, dialog, popup

UserName

H3COHHO

CAS RN 18979-61-8

IUPAC Name: 4-butylbenzene-1,3-diol
Synonyms: 4-butylresorcinol
Molecular Weight (g/mol): 166.22
Molecular Formula: C10H14O2
InChi Key: CSHZYWUPJWVTMQ-UHFFFAOYSA-N
SMILES: CCCCC1=CC=C(O)C=C1O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results