accessibility menu, dialog, popup

UserName

CAS RN 18979-55-0

H3COOH

CAS RN 18979-55-0

IUPAC Name: 4-(hexyloxy)phenol
Synonyms: 4-hexyloxyphenol
Molecular Weight (g/mol): 194.27
Molecular Formula: C12H18O2
InChi Key: XIIIHRLCKLSYNH-UHFFFAOYSA-N
SMILES: CCCCCCOC1=CC=C(O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results