accessibility menu, dialog, popup

UserName

CAS RN 188589-32-4

IUPAC Name: 1-decyl-3-methyl-1H-imidazol-3-ium bromide
Synonyms: 1-decyl-3-methylimidazolium bromide
Molecular Weight (g/mol): 303.29
Molecular Formula: C14H27BrN2
InChi Key: HOISBTKPPVRFDS-UHFFFAOYSA-M
SMILES: [Br-].CCCCCCCCCCN1C=C[N+](C)=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results