accessibility menu, dialog, popup

UserName

N(E)

CAS RN 1885-38-7

IUPAC Name: (2E)-3-phenylprop-2-enenitrile
Synonyms: cinnamonitrile
Molecular Weight (g/mol): 129.16
Molecular Formula: C9H7N
InChi Key: ZWKNLRXFUTWSOY-QPJJXVBHSA-N
SMILES: N#C\C=C\C1=CC=CC=C1
Boiling Point 
  • (2)
  • (3)
  • (1)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (1)
  • (5)
Quantity 
  • (2)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (2)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (5)
  • (2)
  • (3)
  • (1)
  • (4)