missing translation for 'onlineSavingsMsg'
Learn More

Cinnamonitrile 95.0+%, TCI America™

Catalog No. C036125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C036125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C036125G Supplier TCI America Supplier No. C036125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1885-38-7
Molecular Formula C9H7N
Molecular Weight (g/mol) 129.162
MDL Number MFCD00001930
InChI Key ZWKNLRXFUTWSOY-QPJJXVBHSA-N
Synonym cinnamonitrile, trans-cinnamonitrile, styryl cyanide, cinnamyl nitrile, 3-phenylacrylonitrile, beta-cyanostyrene, cinnamonitrile, e, 3-phenyl-2-propenenitrile, acrylonitrile, 3-phenyl, e-3-phenylprop-2-enenitrile
PubChem CID 1550846
IUPAC Name (E)-3-phenylprop-2-enenitrile
SMILES C1=CC=C(C=C1)C=CC#N

Specifications

Color Yellow
Boiling Point 255°C
Quantity 25 g
Formula Weight 129.16
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Cinnamonitrile
EINECSNumber (3)-1814
TSCA Yes