missing translation for 'onlineSavingsMsg'
Learn More

Cinnamonitrile, 97%, predominantly trans

Catalog No. AAB2550322
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2550322 100 g
AAB2550314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2550322 Supplier Thermo Scientific Chemicals Supplier No. B2550322
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1885-38-7 | C9H7N | 129.162 g/mol

Cinnamonitrile is a parent compound whose derivatives can be applied as corrosion inhibitors for API J55 steel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cinnamonitrile is a parent compound whose derivatives can be applied as corrosion inhibitors for API J55 steel.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Air & light sensitive. Store in dark. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 1885-38-7
Molecular Formula C9H7N
Molecular Weight (g/mol) 129.162
MDL Number MFCD00001930
InChI Key ZWKNLRXFUTWSOY-QPJJXVBHSA-N
Synonym cinnamonitrile, trans-cinnamonitrile, styryl cyanide, cinnamyl nitrile, 3-phenylacrylonitrile, beta-cyanostyrene, cinnamonitrile, e, 3-phenyl-2-propenenitrile, acrylonitrile, 3-phenyl, e-3-phenylprop-2-enenitrile
PubChem CID 1550846
IUPAC Name (E)-3-phenylprop-2-enenitrile
SMILES C1=CC=C(C=C1)C=CC#N

Specifications

Melting Point 18°C to 20°C
Density 1.029
Boiling Point 253°C to 254°C
Flash Point 126°C (259°F)
Refractive Index 1.602
Quantity 100 g
Sensitivity Air and light sensitive
Solubility Information Soluble in alcohol. Insoluble in water.
Formula Weight 129.16
Percent Purity 97%
Chemical Name or Material Cinnamonitrile, predominantly trans
Show More Show Less
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 217-552-5
RTECSNumber UD1440000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only