accessibility menu, dialog, popup

UserName

BrSO

CAS RN 18791-75-8

IUPAC Name: 4-bromothiophene-2-carbaldehyde
Synonyms: 4-bromothiophene-2-carbaldehyde
Molecular Weight (g/mol): 191.04
Molecular Formula: C5H3BrOS
InChi Key: PDONIKHDXYHTLS-UHFFFAOYSA-N
SMILES: BrC1=CSC(C=O)=C1
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)