accessibility menu, dialog, popup

UserName

CAS RN 1878-84-8

IUPAC Name: 2-(4-hydroxyphenoxy)acetate
Synonyms: 2-(4-hydroxyphenoxy)acetate
Molecular Weight (g/mol): 167.14
Molecular Formula: C8H7O4
InChi Key: PKGWLCZTTHWKIZ-UHFFFAOYSA-M
SMILES: OC1=CC=C(OCC([O-])=O)C=C1
Molecular Weight (g/mol) 
  • (5)
  • (1)
Percent Purity 
  • (3)
  • (2)
  • (1)
Quantity 
  • (1)
  • (3)
  • (2)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (5)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)