CAS RN 187604-75-7
IUPAC Name:
6-bromo-3,4-dihydro-2H-1,4-benzothiazine
Synonyms:
6-bromo-3,4-dihydro-2H-1,4-benzothiazine
Molecular Weight (g/mol):
230.12
Molecular Formula:
C8H8BrNS
InChi Key:
AUIBYERXYVUBIL-UHFFFAOYSA-N
SMILES:
BrC1=CC2=C(SCCN2)C=C1
1
–
1
of
1
results
1
Promotions Available
6-Bromo-3,4-dihydro-2H-1,4-benzothiazine, 97%
6-Bromo-3,4-dihydro-2H-1,4-benzothiazine, 97%
CAS: 187604-75-7 Molecular Formula: C8H8BrNS Molecular Weight (g/mol): 230.123 MDL Number: MFCD23379863 InChI Key: AUIBYERXYVUBIL-UHFFFAOYSA-N Synonym: 6-bromo-3,4-dihydro-2h-benzo b 1,4 thiazine PubChem CID: 67172430 IUPAC Name: 6-bromo-3,4-dihydro-2H-1,4-benzothiazine SMILES: C1CSC2=C(N1)C=C(C=C2)Br