accessibility menu, dialog, popup

UserName

CH3H3CH3CCH3CH3NH2H2NCH3

CAS RN 186769-18-6

IUPAC Name: 1,2-bis(2,4,6-trimethylphenyl)ethane-1,2-diamine
Synonyms: 1,2-bis(2,4,6-trimethylphenyl)ethane-1,2-diamine
Molecular Weight (g/mol): 296.46
Molecular Formula: C20H28N2
InChi Key: ILMRHFMYIXTNMC-UHFFFAOYNA-N
SMILES: CC1=CC(C)=C(C(N)C(N)C2=C(C)C=C(C)C=C2C)C(C)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results