accessibility menu, dialog, popup

UserName

CAS RN 186590-36-3

IUPAC Name: 4,5-difluoro-2-hydroxybenzonitrile
Synonyms: 4,5-difluoro-2-hydroxybenzonitrile
Molecular Weight (g/mol): 155.1
Molecular Formula: C7H3F2NO
InChi Key: ROLMZTIHUMKEAI-UHFFFAOYSA-N
SMILES: OC1=CC(F)=C(F)C=C1C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results