accessibility menu, dialog, popup

UserName

CAS RN 1860-99-7

IUPAC Name: 2-bromothiophene-3-carbaldehyde
Synonyms: 2-bromothiophene-3-carbaldehyde
Molecular Weight (g/mol): 191.04
Molecular Formula: C5H3BrOS
InChi Key: MMEGVQIGIBCTHI-UHFFFAOYSA-N
SMILES: BrC1=C(C=O)C=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results