accessibility menu, dialog, popup

UserName

CAS RN 1860-58-8

OOOH

CAS RN 1860-58-8

IUPAC Name: 2-[3-(benzyloxy)phenyl]acetic acid
Synonyms: [3-(benzyloxy)phenyl]acetic acid
Molecular Weight (g/mol): 242.27
Molecular Formula: C15H14O3
InChi Key: LLZKAZNUCYYBQO-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC(OCC2=CC=CC=C2)=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results