accessibility menu, dialog, popup

UserName

CAS RN 185346-79-6

IUPAC Name: 4-(benzyloxy)-1-bromo-2-fluorobenzene
Synonyms: 4-(benzyloxy)-1-bromo-2-fluorobenzene
Molecular Weight (g/mol): 281.12
Molecular Formula: C13H10BrFO
InChi Key: BYTJTXKQBSNGCL-UHFFFAOYSA-N
SMILES: FC1=C(Br)C=CC(OCC2=CC=CC=C2)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results