accessibility menu, dialog, popup

UserName

CAS RN 18437-78-0

IUPAC Name: tris(4-fluorophenyl)phosphane
Synonyms: tris(4-fluorophenyl)phosphane
Molecular Weight (g/mol): 316.26
Molecular Formula: C18H12F3P
InChi Key: GEPJPYNDFSOARB-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)P(C1=CC=C(F)C=C1)C1=CC=C(F)C=C1
Percent Purity 
  • (1)
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)
  • (1)