accessibility menu, dialog, popup

UserName

CH3CH2SiH3CCH3CH3

CAS RN 18387-60-5

IUPAC Name: trimethyl(3-methylbut-3-en-1-yn-1-yl)silane
Synonyms: trimethyl(3-methylbut-3-en-1-yn-1-yl)silane
Molecular Weight (g/mol): 138.29
Molecular Formula: C8H14Si
InChi Key: HRGBALJHGYAWBL-UHFFFAOYSA-N
SMILES: CC(=C)C#C[Si](C)(C)C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results