accessibility menu, dialog, popup

UserName

FFNFFN

CAS RN 1835-49-0

IUPAC Name: tetrafluorobenzene-1,4-dicarbonitrile
Synonyms: tetrafluoroterephthalonitrile
Molecular Weight (g/mol): 200.1
Molecular Formula: C8F4N2
InChi Key: PCRSJGWFEMHHEW-UHFFFAOYSA-N
SMILES: FC1=C(F)C(C#N)=C(F)C(F)=C1C#N
Percent Purity 
  • (3)
  • (1)
Physical Form 
  • (3)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)