accessibility menu, dialog, popup

UserName

CAS RN 18344-51-9

CH3HNH2NNOO

CAS RN 18344-51-9

IUPAC Name: 2-amino-3-methyl-5-nitropyridin-1-ium
Synonyms: 2-amino-3-methyl-5-nitropyridin-1-ium
Molecular Weight (g/mol): 154.15
Molecular Formula: C6H8N3O2
InChi Key: JLPYUDJJBGYXEZ-UHFFFAOYSA-O
SMILES: CC1=CC(=C[NH+]=C1N)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results