accessibility menu, dialog, popup

UserName

CAS RN 183438-24-6

IUPAC Name: 5-bromo-2-iodopyrimidine
Synonyms: 5-bromo-2-iodopyrimidine
Molecular Weight (g/mol): 284.88
Molecular Formula: C4H2BrIN2
InChi Key: ZEZKXPQIDURFKA-UHFFFAOYSA-N
SMILES: BrC1=CN=C(I)N=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results