accessibility menu, dialog, popup

UserName

CAS RN 1829-60-3

IUPAC Name: 2,3-dimethyl 7-oxabicyclo[2.2.1]hepta-2,5-diene-2,3-dicarboxylate
Synonyms: 2,3-dimethyl 7-oxabicyclo[2.2.1]hepta-2,5-diene-2,3-dicarboxylate
Molecular Weight (g/mol): 210.19
Molecular Formula: C10H10O5
InChi Key: FQVSHUVBNGSEJO-UHFFFAOYNA-N
SMILES: COC(=O)C1=C(C2OC1C=C2)C(=O)OC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results