accessibility menu, dialog, popup

UserName

NHNOFFFOHNFFF

CAS RN 182431-12-5

IUPAC Name: N-(2,2,2-trifluoroethyl)-9-(4-{4-[4'-(trifluoromethyl)-[1,1'-biphenyl]-2-amido]piperidin-1-yl}butyl)-9H-fluorene-9-carboxamide
Synonyms: lomitapide
Molecular Weight (g/mol): 693.73
Molecular Formula: C39H37F6N3O2
InChi Key: MBBCVAKAJPKAKM-UHFFFAOYSA-N
SMILES: FC(F)(F)CNC(=O)C1(CCCCN2CCC(CC2)NC(=O)C2=CC=CC=C2C2=CC=C(C=C2)C(F)(F)F)C2=CC=CC=C2C2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)