missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lomitapide 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 142°C |
| Color | White |
| Quantity | 25 mg |
| Formula Weight | 693.73 |
| Percent Purity | ≥98.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Lomitapide |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |