accessibility menu, dialog, popup

UserName

CAS RN 18115-70-3

IUPAC Name: lithium(1+) (2Z)-4-oxopent-2-en-2-olate
Synonyms: lithium(1+) (2Z)-4-oxopent-2-en-2-olate
Molecular Weight (g/mol): 106.05
Molecular Formula: C5H7LiO2
InChi Key: JTEOOCAGEXVCBQ-LNKPDPKZSA-M
SMILES: [Li+].C\C([O-])=C\C(C)=O
Percent Purity 
  • (1)
  • (3)
Physical Form 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)