accessibility menu, dialog, popup

UserName

CAS RN 18108-68-4

IUPAC Name: magnesium(2+) bis(8-(phenylamino)naphthalene-1-sulfonate)
Synonyms: magnesium(2+) bis(8-(phenylamino)naphthalene-1-sulfonate)
Molecular Weight (g/mol): 620.98
Molecular Formula: C32H24MgN2O6S2
InChi Key: IRKGXNAZPUOEMW-UHFFFAOYSA-L
SMILES: [Mg++].[O-]S(=O)(=O)C1=C2C(NC3=CC=CC=C3)=CC=CC2=CC=C1.[O-]S(=O)(=O)C1=C2C(NC3=CC=CC=C3)=CC=CC2=CC=C1
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)