accessibility menu, dialog, popup

UserName

H3COPOOCH3

CAS RN 1809-19-4

IUPAC Name: dibutoxy(oxo)-λ⁵-phosphanylium
Synonyms: dibutoxy(oxo)-λ⁵-phosphanylium
Molecular Weight (g/mol): 193.2
Molecular Formula: C8H18O3P
InChi Key: OSPSWZSRKYCQPF-UHFFFAOYSA-N
SMILES: CCCCO[P+](=O)OCCCC
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)