accessibility menu, dialog, popup

UserName

CH3OCH3OCH3OH3COCH3OH3CONNTb(E/Z)(E/Z)(E/Z)

CAS RN 18078-86-9

IUPAC Name: terbium(3+) 1,10-phenanthroline tris(4-oxopent-2-en-2-olate)
Synonyms: terbium(3+) phen tris(4-oxopent-2-en-2-olate)
Molecular Weight (g/mol): 636.46
Molecular Formula: C27H29N2O6Tb
InChi Key: YUDOUPKNKVFAMM-UHFFFAOYSA-K
SMILES: [Tb+3].CC([O-])=CC(C)=O.CC([O-])=CC(C)=O.CC([O-])=CC(C)=O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results