accessibility menu, dialog, popup

UserName

CAS RN 17927-72-9

TiH3CH3COCH3CH3OCH3OCHCH3OCH3OCHCH3O

CAS RN 17927-72-9

IUPAC Name: titanium(4+) bis(2,4-dioxopentan-3-ide) bis(propan-2-olate)
Synonyms: titanium(4+) diacetylacetonate bis(propan-2-olate)
Molecular Weight (g/mol): 364.26
Molecular Formula: C16H28O6Ti
InChi Key: DOOBMVWXHWRGEB-UHFFFAOYSA-N
SMILES: [Ti+4].CC(C)[O-].CC(C)[O-].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results