accessibility menu, dialog, popup

UserName

CH3NOSOOFFF

CAS RN 179260-78-7

IUPAC Name: 4-methylpyridin-2-yl trifluoromethanesulfonate
Synonyms: 4-methylpyridin-2-yl trifluoromethanesulfonate
Molecular Weight (g/mol): 241.18
Molecular Formula: C7H6F3NO3S
InChi Key: AZUJRVRRUXHHDV-UHFFFAOYSA-N
SMILES: CC1=CC(OS(=O)(=O)C(F)(F)F)=NC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results