accessibility menu, dialog, popup

UserName

POOHBr

CAS RN 17857-14-6

IUPAC Name: (3-carboxypropyl)triphenylphosphanium bromide
Synonyms: (3-carboxypropyl)triphenylphosphanium bromide
Molecular Weight (g/mol): 429.29
Molecular Formula: C22H22BrO2P
InChi Key: NKVJKVMGJABKHV-UHFFFAOYSA-N
SMILES: [Br-].OC(=O)CCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
  • (4)
Quantity 
  • (2)
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (1)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)