missing translation for 'onlineSavingsMsg'
Learn More

(3-Carboxypropyl)triphenylphosphonium bromide, 97%

Catalog No. AAA1930236
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1930236 500 g
AAA1930214 25 g
AAA1930222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1930236 Supplier Thermo Scientific Chemicals Supplier No. A1930236
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 17857-14-6 | C22H22BrO2P | 429.29 g/mol

Reactant for preparation of Piperamide analogs as histone deacetylase (HDAC) inhibitors with antitumor activity, Boron-containing benzoxaboroles as antimalarial agents, Methyl alkenyl quinolones as antimycobacterial agents, Solandelactone E via Sharpless epoxidation, Taber cyclopropanation, chemoselective reductions and lithiation-borylation-allylation sequence, Organotin compounds as antifungal agents, Oleanolic acid derivatives as inhibitors of protein tyrosine phosphatase 1B for negative regulation of insulin pathway (a promising target for treatment of diabetes and obesity), 11-oxa prostaglandin analogs with ocular hypotensive activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant for preparation of Piperamide analogs as histone deacetylase (HDAC) inhibitors with antitumor activity, Boron-containing benzoxaboroles as antimalarial agents, Methyl alkenyl quinolones as antimycobacterial agents, Solandelactone E via Sharpless epoxidation, Taber cyclopropanation, chemoselective reductions and lithiation-borylation-allylation sequence, Organotin compounds as antifungal agents, Oleanolic acid derivatives as inhibitors of protein tyrosine phosphatase 1B for negative regulation of insulin pathway (a promising target for treatment of diabetes and obesity), 11-oxa prostaglandin analogs with ocular hypotensive activity.

Solubility
Soluble in water. soluble in ethanol.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.

Chemical Identifiers

CAS 17857-14-6
Molecular Formula C22H22BrO2P
Molecular Weight (g/mol) 429.29
MDL Number MFCD00274196
InChI Key NKVJKVMGJABKHV-UHFFFAOYSA-N
Synonym 3-carboxypropyltriphenylphosphonium bromide, 3-carboxypropyl triphenylphosphonium bromide, 3-carboxypropyl triphenylphosphanium bromide, carboxypropyltriphenylphosphoniumbromide, phosphonium, 3-carboxypropyl triphenyl-, bromide, 4-hydroxy-4-oxobutyl triphenylphosphonium bromide, 3-carboxypropyl triphenylphosphonium bromide 3-carboxypropyl triphenylphosphonium bromide, acmc-1bp1u, xfzdcviyhhhybq-uhfffaoysa-m
PubChem CID 10717451
IUPAC Name 3-carboxypropyl(triphenyl)phosphanium;bromide
SMILES [Br-].OC(=O)CCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 244°C to 250°C
Quantity 500 g
Beilstein 4173599
Sensitivity Hygroscopic
Solubility Information Soluble in water. soluble in ethanol.
Formula Weight 429.3
Percent Purity 97%
Chemical Name or Material (3-Carboxypropyl)triphenylphosphonium bromide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only