missing translation for 'onlineSavingsMsg'
Learn More

(3-Carboxypropyl)triphenylphosphonium Bromide 95.0+%, TCI America™

Catalog No. C163525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C163525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C163525G Supplier TCI America Supplier No. C163525G

Chemical Identifiers

CAS 17857-14-6
Molecular Formula C22H22BrO2P
Molecular Weight (g/mol) 429.29
MDL Number MFCD00274196
InChI Key NKVJKVMGJABKHV-UHFFFAOYSA-N
Synonym 3-carboxypropyltriphenylphosphonium bromide, 3-carboxypropyl triphenylphosphonium bromide, 3-carboxypropyl triphenylphosphanium bromide, carboxypropyltriphenylphosphoniumbromide, phosphonium, 3-carboxypropyl triphenyl-, bromide, 4-hydroxy-4-oxobutyl triphenylphosphonium bromide, 3-carboxypropyl triphenylphosphonium bromide 3-carboxypropyl triphenylphosphonium bromide, acmc-1bp1u, xfzdcviyhhhybq-uhfffaoysa-m
PubChem CID 10717451
IUPAC Name (3-carboxypropyl)triphenylphosphanium bromide
SMILES [Br-].OC(=O)CCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 248°C
Quantity 25 g
Formula Weight 429.29
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (3-Carboxypropyl)triphenylphosphonium Bromide