accessibility menu, dialog, popup

UserName

FNNF

CAS RN 178439-26-4

IUPAC Name: 1,1'-difluoro-[2,2'-bipyridine]-1,1'-diium
Synonyms: 1,1'-difluoro-[2,2'-bipyridine]-1,1'-diium
Molecular Weight (g/mol): 194.18
Molecular Formula: C10H8F2N2
InChi Key: VWXNMFVNVKSKOB-UHFFFAOYSA-N
SMILES: F[N+]1=CC=CC=C1C1=CC=CC=[N+]1F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results