accessibility menu, dialog, popup

UserName

BrHOOP

CAS RN 17814-85-6

IUPAC Name: (4-carboxybutyl)triphenylphosphanium bromide
Synonyms: (4-carboxybutyl)triphenylphosphanium bromide
Molecular Weight (g/mol): 443.32
Molecular Formula: C23H24BrO2P
InChi Key: MLOSJPZSZWUDSK-UHFFFAOYSA-N
SMILES: [Br-].OC(=O)CCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)