missing translation for 'onlineSavingsMsg'
Learn More

4-(Carboxybutyl)triphenylphosphonium Bromide 98.0+%, TCI America™

Catalog No. C106125G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C106125G 25 g
C1061250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C106125G Supplier TCI America Supplier No. C106125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 17814-85-6
Molecular Formula C23H24BrO2P
Molecular Weight (g/mol) 443.32
MDL Number MFCD00011906
InChI Key MLOSJPZSZWUDSK-UHFFFAOYSA-N
Synonym 4-carboxybutyl triphenylphosphonium bromide, phosphonium, 4-carboxybutyl triphenyl-, bromide, 4-carboxybutyl triphenylphosphanium bromide, carboxybutyltriphenylphosphonium bromide, 5-triphenylphosphonio pentanoic acid bromide, 4-carboxybutyl-triphenylphosphonium bromide, triphenyl 4-carboxybutyl phosphonium bromide, 4-carboxybutyl triphenyl phosphonium bromide
PubChem CID 161236
IUPAC Name (4-carboxybutyl)triphenylphosphanium bromide
SMILES [Br-].OC(=O)CCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 207°C
Color White
Quantity 25 g
Formula Weight 443.32
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Carboxybutyl)triphenylphosphonium Bromide
EINECSNumber (3)-3356
TSCA No