missing translation for 'onlineSavingsMsg'
Learn More

(4-Carboxybutyl)triphenylphosphonium bromide, 98%

Catalog No. AAA1202336
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1202336 500 g
AAA1202314 25 g
AAA1202322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1202336 Supplier Thermo Scientific Chemicals Supplier No. A1202336
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 17814-85-6 | C23H24BrO2P | 443.32 g/mol

Phosphonium salt is a key intermediate for manufacture of some types of prostaglandins, which are subsequently used as active ingredients of both human and veterinary drugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phosphonium salt is a key intermediate for manufacture of some types of prostaglandins, which are subsequently used as active ingredients of both human and veterinary drugs.

Solubility
Soluble in ethanol, methanol and soluble water. Insoluble in toluene and hexane.

Notes
Hygroscopic. Store in dry place. Keep away from oxidizing agents.

Chemical Identifiers

CAS 17814-85-6
Molecular Formula C23H24BrO2P
Molecular Weight (g/mol) 443.32
MDL Number MFCD00011906
InChI Key MLOSJPZSZWUDSK-UHFFFAOYSA-N
Synonym 4-carboxybutyl triphenylphosphonium bromide, phosphonium, 4-carboxybutyl triphenyl-, bromide, 4-carboxybutyl triphenylphosphanium bromide, carboxybutyltriphenylphosphonium bromide, 5-triphenylphosphonio pentanoic acid bromide, 4-carboxybutyl-triphenylphosphonium bromide, triphenyl 4-carboxybutyl phosphonium bromide, 4-carboxybutyl triphenyl phosphonium bromide
PubChem CID 161236
IUPAC Name 4-carboxybutyl(triphenyl)phosphanium;bromide
SMILES [Br-].OC(=O)CCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 205°C to 209°C
Flash Point 195°C (383°F)
Linear Formula HO2C(CH2)4P(C6H5)3Br
Quantity 500 g
Beilstein 3586477
Sensitivity Hygroscopic
Solubility Information Soluble in ethanol,methanol and soluble water. Insoluble in toluene and hexane.
Formula Weight 443.33
Percent Purity 98%
Chemical Name or Material (4-Carboxybutyl)triphenylphosphonium bromide
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 241-782-5
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only