accessibility menu, dialog, popup

UserName

CAS RN 17796-82-6

ONSO

CAS RN 17796-82-6

IUPAC Name: 2-(cyclohexylsulfanyl)-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: N-(cyclohexylthio)phthalimide cyclohexylthiophthalimide
Molecular Weight (g/mol): 261.34
Molecular Formula: C14H15NO2S
InChi Key: UEZWYKZHXASYJN-UHFFFAOYSA-N
SMILES: O=C1N(SC2CCCCC2)C(=O)C2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results