accessibility menu, dialog, popup

UserName

ONOClClCl

CAS RN 17700-09-3

IUPAC Name: 1,2,3-trichloro-4-nitrobenzene
Synonyms: 2,3,4-trichloronitrobenzene
Molecular Weight (g/mol): 226.44
Molecular Formula: C6H2Cl3NO2
InChi Key: BGKIECJVXXHLDP-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(Cl)C(Cl)=C1Cl
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results