accessibility menu, dialog, popup

UserName

CH3SN

CAS RN 17626-75-4

IUPAC Name: 2-propyl-1,3-thiazole
Synonyms: 2-propyl-1,3-thiazole
Molecular Weight (g/mol): 127.21
Molecular Formula: C6H9NS
InChi Key: CMOIEFFAOUQJPS-UHFFFAOYSA-N
SMILES: CCCC1=NC=CS1
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)