accessibility menu, dialog, popup

UserName

ClH3CCH3(S)NH2OOOHONNONNH2HNH

CAS RN 175865-59-5

IUPAC Name: hydrogen 2-[(2-amino-6-oxo-6,9-dihydro-3H-purin-9-yl)methoxy]-3-hydroxypropyl (2S)-2-amino-3-methylbutanoate chloride
Synonyms: valganciclovir
Molecular Weight (g/mol): 390.83
Molecular Formula: C14H23ClN6O5
InChi Key: ZORWARFPXPVJLW-MTFPJWTKSA-N
SMILES: [H+].[Cl-].CC(C)[C@H](N)C(=O)OCC(CO)OCN1C=NC2=C1NC(N)=NC2=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)