CAS RN 17557-23-2
CAS RN 17557-23-2
IUPAC Name:
2-({2,2-dimethyl-3-[(oxiran-2-yl)methoxy]propoxy}methyl)oxirane
Synonyms:
neopentyl glycol diglycidyl ether
Molecular Weight (g/mol):
216.28
Molecular Formula:
C11H20O4
InChi Key:
KUAUJXBLDYVELT-UHFFFAOYNA-N
SMILES:
CC(C)(COCC1CO1)COCC1CO1
1
–
1
of
1
results
1
Neopentyl Glycol Diglycidyl Ether 40.0+%, TCI America™
CAS: 17557-23-2 Molecular Formula: C11H20O4 Molecular Weight (g/mol): 216.277 MDL Number: MFCD00040687 InChI Key: KUAUJXBLDYVELT-UHFFFAOYSA-N Synonym: Diglycidyloxyneopentane PubChem CID: 28594 IUPAC Name: 2-[[2,2-dimethyl-3-(oxiran-2-ylmethoxy)propoxy]methyl]oxirane SMILES: CC(C)(COCC1CO1)COCC2CO2