CAS RN 175230-50-9
IUPAC Name:
propan-2-yl 4,4,4-trifluoro-3-oxobutanoate
Synonyms:
isopropyl 4,4,4-trifluoro-3-oxobutanoate
Molecular Weight (g/mol):
198.14
Molecular Formula:
C7H9F3O3
InChi Key:
XLBGYZLICFMDDS-UHFFFAOYSA-N
SMILES:
CC(C)OC(=O)CC(=O)C(F)(F)F
1
–
1
of
1
results
1
Isopropyl 4,4,4-Trifluoroacetoacetate 95.0+%, TCI America™
CAS: 175230-50-9 Molecular Formula: C7H9F3O3 Molecular Weight (g/mol): 198.14 MDL Number: MFCD00040990 InChI Key: XLBGYZLICFMDDS-UHFFFAOYSA-N Synonym: isopropyl 4,4,4-trifluoroacetoacetate,isopropyl 4,4,4-trifluoro-3-oxobutanoate,isopropyl 4,4,4-trifluoro-3-oxobutyrate,isopropyl-4,4,4-trifluoroacetoacetate,acmc-209e9i,isopropyl4,4,4-trifluoroacetoacetate,methylethyl 4,4,4-trifluoro-3-oxobutanoate,isopropyl 4,4,4-trifluoro-3-oxobutanoate #,4,4,4-trifluoroacetoacetic acid isopropyl ester PubChem CID: 586897 IUPAC Name: propan-2-yl 4,4,4-trifluoro-3-oxobutanoate SMILES: CC(C)OC(=O)CC(=O)C(F)(F)F