accessibility menu, dialog, popup

UserName

CH3NNH3CFFFSOOO

CAS RN 174899-66-2

IUPAC Name: 1-butyl-3-methyl-1H-imidazol-3-ium trifluoromethanesulfonate
Synonyms: 1-butyl-3-methylimidazolium triflate
Molecular Weight (g/mol): 288.29
Molecular Formula: C9H15F3N2O3S
InChi Key: FRZPYEHDSAQGAS-UHFFFAOYSA-M
SMILES: [O-]S(=O)(=O)C(F)(F)F.CCCCN1C=C[N+](C)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)