accessibility menu, dialog, popup

UserName

CAS RN 174813-81-1

CH3SOONHNH2CH3H3CCH3ClRu

CAS RN 174813-81-1

IUPAC Name: λ¹-ruthenium(1+) 1,3,5-trimethylbenzene N-(2-amino-1,2-diphenylethyl)-4-methylbenzene-1-sulfonamide chloride
Synonyms: λ¹-ruthenium(1+) N-(2-amino-1,2-diphenylethyl)-4-methylbenzenesulfonamide mesitylene chloride
Molecular Weight (g/mol): 623.19
Molecular Formula: C30H34ClN2O2RuS
InChi Key: BABSEZBJIPZYQB-UHFFFAOYNA-M
SMILES: [Cl-].[Ru+].CC1=CC(C)=CC(C)=C1.CC1=CC=C(C=C1)S(=O)(=O)NC(C(N)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results