CAS RN 174403-69-1
IUPAC Name:
methyl 4-fluoro-2-methylbenzoate
Synonyms:
methyl 4-fluoro-2-methylbenzoate
Molecular Weight (g/mol):
168.17
Molecular Formula:
C9H9FO2
InChi Key:
ZQFCTCYDRQFPBU-UHFFFAOYSA-N
SMILES:
COC(=O)C1=C(C)C=C(F)C=C1
1
–
1
of
1
results
1
Promotions Available
Methyl 4-fluoro-2-methylbenzoate, 98%
Methyl 4-fluoro-2-methylbenzoate, 98%
CAS: 174403-69-1 Molecular Formula: C9H9FO2 Molecular Weight (g/mol): 168.17 MDL Number: MFCD06203790 InChI Key: ZQFCTCYDRQFPBU-UHFFFAOYSA-N Synonym: benzoic acid, 4-fluoro-2-methyl-, methyl ester,4-fluoro-2-methylbenzoic acid methyl ester,rarechem al bf 0506,methyl=4-fluoro-2-methylbenzoate,4-fluoro-2-methyl benzoic acid methyl ester,4-fluoro-2-methyl-benzoic acid methyl ester,benzoic acid,4-fluoro-2-methyl-,methyl ester PubChem CID: 2782181 IUPAC Name: methyl 4-fluoro-2-methylbenzoate SMILES: COC(=O)C1=C(C)C=C(F)C=C1