accessibility menu, dialog, popup

UserName

NH2NNOO

CAS RN 17420-30-3

IUPAC Name: 2-amino-5-nitrobenzonitrile
Synonyms: benzonitrile, 2-amino-5-nitro-
Molecular Weight (g/mol): 163.14
Molecular Formula: C7H5N3O2
InChi Key: MGCGMYPNXAFGFA-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1C#N)[N+]([O-])=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)