CAS RN 17371-99-2
CAS RN 17371-99-2
IUPAC Name:
5-phenyl-2,3-dihydro-1,3-benzoxazole-2-thione
Synonyms:
5-phenyl-3H-1,3-benzoxazole-2-thione
Molecular Weight (g/mol):
227.28
Molecular Formula:
C13H9NOS
InChi Key:
QKLMWNVNJBXKRG-UHFFFAOYSA-N
SMILES:
S=C1NC2=CC(=CC=C2O1)C1=CC=CC=C1
1
–
1
of
1
results
1
5-Phenylbenzoxazole-2-thiol 98.0+%, TCI America™
CAS: 17371-99-2 Molecular Formula: C13H9NOS Molecular Weight (g/mol): 227.28 MDL Number: MFCD00022847 InChI Key: QKLMWNVNJBXKRG-UHFFFAOYSA-N Synonym: 2-Mercapto-5-phenylbenzoxazole, 5-Phenylbenzoxazoline-2(3H)-thione PubChem CID: 3032663 IUPAC Name: 5-phenyl-2,3-dihydro-1,3-benzoxazole-2-thione SMILES: S=C1NC2=CC(=CC=C2O1)C1=CC=CC=C1