accessibility menu, dialog, popup

UserName

SCNZnBr

CAS RN 173382-28-0

IUPAC Name: 1,3-thiazol-2-ide; bromozincylium
Synonyms: 1,3-thiazol-2-ide; bromozincylium
Molecular Weight (g/mol): 229.4
Molecular Formula: C3H2BrNSZn
InChi Key: YORIBCPQDAVKHG-UHFFFAOYSA-M
SMILES: [Zn+]Br.S1C=CN=[C-]1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)