accessibility menu, dialog, popup

UserName

HOOP

CAS RN 17261-28-8

IUPAC Name: 2-(diphenylphosphanyl)benzoic acid
Synonyms: 2-(diphenylphosphanyl)benzoic acid
Molecular Weight (g/mol): 306.3
Molecular Formula: C19H15O2P
InChi Key: UYRPRYSDOVYCOU-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)