accessibility menu, dialog, popup

UserName

CAS RN 17260-71-8

IUPAC Name: 3-chlorobenzene-1-sulfonamide
Synonyms: M-chlorobenzenesulfonamide
Molecular Weight (g/mol): 191.63
Molecular Formula: C6H6ClNO2S
InChi Key: WSYQJNPRQUFCGL-UHFFFAOYSA-N
SMILES: NS(=O)(=O)C1=CC=CC(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results